Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-B220/CD45R, Alexa Fluor 750, Clone: RA3-6B2, R&D Systems™
Rat Monoclonal Antibody
Specifications
Antigen | B220/CD45R |
---|---|
Clone | RA3-6B2 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1217S025
|
R&D Systems
FAB1217S025 |
25 Tests | N/A | N/A | N/A | ||||
FAB1217S100
|
R&D Systems
FAB1217S100 |
100 Tests | N/A | N/A | N/A | ||||
Specifications
B220/CD45R | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Supernatant | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
5788 | |
Abelson mouse leukemia virus-induced pre-B tumor cells | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
RA3-6B2 | |
Flow Cytometry | |
Alexa Fluor 750 | |
Rat | |
Mouse | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
PTPRC | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse B220/CD45R. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title