Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD57 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526921
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7).
Specifications
CD57 | |
Polyclonal | |
B3GAT1 | |
CD57, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1, LEU7 antigen, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, glcUAT-P, glucuronosyltransferase P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27087 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q9P2W7 | |
B3GAT1 | |
KLH conjugated synthetic peptide between 21-48 amino acids from the N-terminal region of human B3GAT1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction