Learn More
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant B3GNT3.
Supplier: Abnova Corporation H00010331A01
Description
This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein and contains a signal anchor that is not cleaved. It prefers the substrates of lacto-N-tetraose and lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains and the biosynthesis of the backbone structure of dimeric sialyl Lewis a. It plays dominant roles in L-selectin ligand biosynthesis, lymphocyte homing and lymphocyte trafficking. [provided by RefSeq
Sequence: KVRGLQLRLLFLVGTASNPHEARKVNRLLELEAQTHGDILQWDFHDSFFNLTLKQVLFLQWQETRCANASFVLNSpecifications
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant B3GNT3. | |
B3GNT3 | |
B3GAL-T8/B3GN-T3/B3GNT-3/HP10328/TMEM3/beta3Gn-T3 | |
Mouse | |
50 μL | |
Yes | |
10331 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_014256 | |
B3GNT3 | |
B3GNT3 (NP_055071, 135 a.a. ∼ 208 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.