Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B3GNT6 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526883
Description

Specifications
B3GNT6 | |
Polyclonal | |
B3GNT6 | |
B3Gn-T6, IMAGE:4907098, BGnT-6, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6, beta-1,3-Gn-T6, beta-1,3-N-acetylglucosaminyltransferase 6, beta-1,3-N-acetylglucosaminyltransferase protein, beta3Gn-T6, core 3 synthase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
192134, 272411 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Q3USF0, Q6ZMB0 | |
B3GNT6 | |
KLH conjugated synthetic peptide between 146-173 amino acids from the central region of human B3GNT6 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction