Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B56α Mouse, Unlabeled, Clone: 23, BD

Catalog No. BDB610615 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610615 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610615 Supplier BD Biosciences Supplier No. 610615
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reversible phosphorylation by protein kinases and phosphatases regulates key signaling pathways in eukaryotic cells. Type 2A protein phosphatases (PP2A) are composed of two regulatory and one catalytic subunit. The active PP2A is a heterotrimer, composed of A, B, and C subunits, each encoded by different genes. B56 is a new gene family that encodes α, β, and γ proteins for the B subunit. The three B56 isoforms are approximately 70% homologous to each other and more divergent within the first 251amino acids. While the α subunit is widely expressed, the γ isoform is most abundant in heart and muscle, and the β subunit is found predominantly in brain tissue. Availability of different B subunits appears to be a key determinant in the protein substrate specificity for PP2A and could influence the subcellular distribution of the holoenzyme.

TRUSTED_SUSTAINABILITY

Specifications

Antigen B56α
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human B56α aa. 1-162
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.