Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H3 (CD276) Rat anti-Human, Mouse, Clone: MJ8, MilliporeSigma™

Catalog No. 041501MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-150-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-150-1MI Supplier MilliporeSigma Supplier No. 041501
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect B7-H3 (CD276) using this Anti-B7-H3 (CD276) Antibody, clone MJ8 validated for use in western blot, flow cytometry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-H3 (CD276)
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MJ8
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_598744.1
Gene Symbols CD276, 4Ig-B7-H3, B7-H3, B7H3
Host Species Rat
Immunogen Mouse B7-H3 Ig2a FC fusion protein, consisting of the extracellular domain linked to the Fc portion of mouse IgG2a.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.