Learn More
B7-H4 Mouse anti-Human, Unlabeled, Clone: MIH43, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562506
Description
The MIH43 monoclonal antibody specifically binds to human B7-H4 (B7 family member, H4) that is encoded by the VTCN1 (V-set domain containing T cell activation inhibitor 1) gene. B7-H4 is a type I membrane glycoprotein with a calculated molecular weight of 30.89 kDa. It is also known as VCTN1, B7h.5, B7S1 (B7 superfamily member 1) or B7X. B7-H4 is a newly discovered member of the B7 family of costimulatory proteins. B7-H4 is not constitutively expressed on peripheral tissues. Its expression can be induced on macrophages, dendritic cells, B cells and T cells. By binding to its putative receptor, the function of B7-H4 was initially reported to be a negative regulator of T cell activation, proliferation and differentiation related to cytokine production and cytotoxic effector cell functions. B7-H4 is reportedly overexpressed in a variety tumors. B7-H4 expression by tumor macrophages appears to play a role in suppression antigen-specific T cell mediated immunity. Recent studies indicate that B7-H4 can also be a positive regulator of T cell responses. B7-H4 can be involved innate immunity as well, eg, by inhibiting neutrophil expansion.

Specifications
B7-H4 | |
Monoclonal | |
0.5mg/mL | |
VTCN1; VCTN1; B7 family member, H4; B7H4; B7h.5; B7S1; B7X | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
MIH43 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant human B7-H4 protein | |
50 μg | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.