Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse BACE-1 Ectodomain Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. p-5316813 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB931SP 25 μg
MAB931100 100 μg
MAB931500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB931SP Supplier R&D Systems Supplier No. MAB931SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

BACE-1 Monoclonal specifically detects BACE-1 in Canine∣Human∣Mouse∣Rat samples. It is validated for CyTOF∣ELISA Capture∣ELISA Detection∣ELISA Development∣Flow Cytometry (Intracellular Staining)∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunoprecipitation∣Western Blot.

Specifications

Antigen BACE-1
Applications CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Development, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 137612
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P56817
Gene Alias APP beta-secretase, ASP2, Aspartyl protease 2, BACE1, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Gene Symbols BACE1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BACE-1 Thr22-Tyr460 Accession # P56817
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23621
Test Specificity Detects human and mouse BACE-1 in direct ELISAs and human BACE-1 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BACE-2, recombinant mouse (rm) BACE-2, rhADAM8, rmADAM9, rmADAM10, rhADAM15, or rhTACE is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Canine, Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.