Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5832MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5832MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5832MI Supplier MilliporeSigma Supplier No. AB5832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-BACE Antibody is an antibody against BACE for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BACE
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid. Buffer = PBS containing 0.02% sodium azide.
Gene Symbols BACE1; ASP2; beta-secretase; KIAA1149; Memapsin-2; BACE; memapsin-2; HSPC104; FLJ90568; EC 3.4.23.46
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 458 to 501 of human BACE (Vassar et al. 1999).
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_012104.3; NM_138971.2; NM_138972.2; NM_138973.2
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.