Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE Mouse anti-Human, Mouse, Rat, Primate, Clone: 61-3E7, MilliporeSigma™

Catalog No. MAB5308MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5308MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5308MI Supplier MilliporeSigma Supplier No. MAB5308
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect BACE using this Anti-BACE Antibody, C-terminus, clone 61-3E7 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BACE
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 61-3E7
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid. Buffer = 0.02M Sodium Phosphate, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_012104.3; NM_138971.2; NM_138972.2; NM_138973.2
Gene Symbols BACE1, ASP2, beta-secretase, KIAA1149, Memapsin-2, BACE, memapsin-2, HSPC104, FLJ90568, EC 3.4.23.46
Host Species Mouse
Immunogen Synthetic peptide corresponding to the C-terminus of human BACE.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Primate, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.