Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacterial Peptidoglycan Mouse anti-Bacteria, Clone: 3F6B3, MilliporeSigma™

Catalog No. MAB995MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB995MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB995MI Supplier MilliporeSigma Supplier No. MAB995

Mouse Monoclonal Antibody

This Anti-Bacterial Peptidoglycan Antibody, clone 3F6B3 is validated for use in DB, ELISA, IC for the detection of Bacterial Peptidoglycan.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bacterial Peptidoglycan
Applications Dot Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3F6B3
Conjugate Unconjugated
Formulation Ascites fluid, no preservatives. Liquid.
Host Species Mouse
Immunogen Insoluble PG obtained by TCA-heat and ethanol extraction of Streptococcus mutans BHT cells.
Quantity 100 μL
Primary or Secondary Primary
Target Species Bacteria
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.