Learn More
Bad Mouse, Unlabeled, Clone: 48, BD
Mouse Monoclonal Antibody
Specifications
Antigen | Bad |
---|---|
Clone | 48 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB610391
|
BD Biosciences
610391 |
50 μg | N/A | N/A | N/A | ||||
Description
Isolated by screening for Bcl-2 interacting proteins, Bad shows significant homology to Bcl-2 within the Bcl-2 homology domains 1 and 2 (BH1 and BH2). In addition, several other proteins involved in cell death such as Bax, Bcl-X[L], Mcl-1, and A1 share similar homology with Bcl-2. Bcl-2 is known to oppose several apoptotic signals and is considered to be a central downstream cell death repressor. Bcl-X[L] represses apoptosis, but its short form, Bcl-X[S], promotes cell death. Bax is known to homodimerize as well as heterodimerize with Bcl-2. An excess concentration of Bax opposes the ability of Bcl-2 to repress cell death. Bad can selectively dimerize with Bcl-X[L] and Bcl-2, but not with Bax, Bcl-X[S], Mcl-1, A1, or itself. In mammalian cells, Bad binds more strongly to Bcl-X[L] than Bcl-2. This may explain why Bad reverses the death repressor activity of Bcl-X[L], but not that of Bcl-2. The formation of the Bad-Bcl-X[L] heterodimer displaces Bax andrestores favorable conditions for apoptosis.
This antibody is tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Bad | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2b | |
Affinity Purified |
48 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.