Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bad Mouse, FITC, Clone: 48, BD

Catalog No. BDB610393 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610393 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610393 Supplier BD Biosciences Supplier No. 610393

Mouse Monoclonal Antibody

Isolated by screening for Bcl-2 interacting proteins, Bad shows significant homology to Bcl-2 within the Bcl-2 homology domains 1 and 2 (BH1 and BH2). In addition, several other proteins involved in cell death such as Bax, Bcl-X[L], Mcl-1, and A1 share similar homology with Bcl-2. Bcl-2 is known to oppose several apoptotic signals and is considered to be a central downstream cell death repressor. Bcl-X[L] represses apoptosis, but its short form, Bcl-X[S], promotes cell death. Bax is known to homodimerize as well as heterodimerize with Bcl-2. An excess concentration of Bax opposes the ability of Bcl-2 to repress cell death. Bad can selectively dimerize with Bcl-X[L] and Bcl-2, but not with Bax, Bcl-X[S], Mcl-1, A1, or itself. In mammalian cells, Bad binds more strongly to Bcl-X[L] than Bcl-2. This may explain why Bad reverses the death repressor activity of Bcl-X[L], but not that of Bcl-2. The formation of the Bad-Bcl-X[L] heterodimer displaces Bax andrestores favorable conditions for apoptosis.

This antibody is routinely tested by immunofluoresence microscopy. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bad
Applications Immunofluorescence
Classification Monoclonal
Clone 48
Concentration 250μg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.