Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAF250a/ARID1a Mouse anti-Human, Clone: PSG3, MilliporeSigma™

Catalog No. 04080MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-080-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-080-MI Supplier MilliporeSigma Supplier No. 04080
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-BAF250a/ARID1a Antibody, clone PSG3 is an antibody against BAF250a/ARID1a for use in western blot, Chimmunoprecipitation, IC and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF250a/ARID1a
Applications ChIP Assay, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone PSG3
Conjugate Unconjugated
Formulation 100μg of Protein A purified mouse monoclonal IgG in 100μL of 1X PBS, pH 7.0, 0.1% Azide.
Gene Accession No. NM_006015.4; NM_139135.2
Gene Symbols ARID1A, BAF250, B120, P270, SMARCF1, BAF250A, C10rf4, C1orf4, OSA1, BAF250a, p270, BM029, hELD, OTTHUMP00000044975, hOSA1
Host Species Mouse
Immunogen GST fusion protein corresponding to 419 residues in the mid-portion of human BAF250a/ARID1a
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.