Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAF57 Rabbit anti-Human, Rat, Mouse, Horse, Rhesus Macaque, Bovine, Polyclonal, MilliporeSigma™

Catalog No. ABD52MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABD52MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABD52MI Supplier MilliporeSigma Supplier No. ABD52

Rabbit Polyclonal Antibody

Use Anti-BAF57 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunohistochemistry (paraffin) to detect BAF57 also known as BRG1-associated factor 57 or BAF57.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF57
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols SMARCE1; BAF57
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human BAF57 at the HMG box region.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003070
Target Species Bovine, Equine, Human, Mouse, Rat, Rhesus Monkey
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.