Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAF60a Mouse, Unlabeled, Clone: 23, BD

Catalog No. BDB611728 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611728 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611728 Supplier BD Biosciences Supplier No. 611728
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SWI/SNF complexes facilitate gene activation and transcription factor binding byaltering repressive chromatin structures. In mammals, SWI/SNF complexes are present inmutliple forms that include 9-12 proteins called B RG1- a ssociated f actors (BAFs), whichrange in molecular weight from 47 to 250 kDa. BAF60 is the homologue of the yeastSWP70 subunit of the yeast SWI/SNF complex that regulates glucocorticoidtranscription. Three isoforms of BAF60 are BAF60a, BAF60b, and BAF60c. BAF60a isubiquitously expressed and is present in the 200-kDa BRG1 complex. BAF60b andBAF60c are expressed preferentially in muscle and pancreas and may be subunits of adistinct complex that shares some subunits with BRG1 complex. In hematopoietic cells,a SWI/SNF related-complex (PYR) contains BAF57, INI1, BAF60a, and BAF170. This complex binds a 250 bp pyrimidine-rich element between the fetal and adult ß-globingenes and may facilitate human fetal-to-adult globin gene switching. Thus, BAF60proteins are components of multiple SWI/SNF complexes that regulate the transcriptionof various genes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF60a
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse BAF60a, aa 1-91
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.