Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFFR/TNFRSF13C Rat anti-Mouse, Clone: 204406, R&D Systems™

Catalog No. MAB1755
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1755 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1755 Supplier R&D Systems Supplier No. MAB1755
This item is not returnable. View return policy

Rat Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry, CyTOF.

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 204406
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9D8D0
Gene Symbols Tnfrsf13c
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse BAFF R/TNFRSF13C Gly2-Ala71 Accession # Q9D8D0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115650
Test Specificity Detects mouse BAFF R/TNFRSF13C in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) BAFF R is observed, approximately 10-25% cross-reactivity with recombinant mouse (rm) 4-1BB, rmDR3, rhDR6, rmEDAR, rhHVEM, rmNGF R, rmOX40, and rmTROY is observed, and no cross-reactivity with rmCD27, rmCD30, rmCD40, rmFAS, rmGITR, rmLymphotoxin beta R, rmOPG, rmRANK, rmTNFRI, or rmTNF RII is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.