Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAG-1 Mouse anti-Human, Clone: 3.10G3E2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC525
Description
This Anti-BAG-1 Antibody, clone 3.10G3E2 is validated for use in Western Blotting and Immunohistochemistry and Immunoprecipitation and ELISA for the detection of BAG-1.
Specifications
BAG-1 | |
Monoclonal | |
Unconjugated | |
NP_001165886 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
3.10G3E2 | |
Purified mouse monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
BAG1, HAP | |
GST-tagged recombinant protein corresponding to Human BAG-1. | |
100 μL | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction