Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAG3 Mouse anti-Human, Clone: AC-1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC282
Description
Detect BAG3 using this Anti-BAG3 Antibody, clone AC-1 validated for use in Immunoprecipitation, immunohistochemistry (paraffin).
Specifications
BAG3 | |
Monoclonal | |
Unconjugated | |
NP_004272 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunohistochemistry, Immunoprecipitation | |
AC-1 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
BAG3, BIS | |
MAP peptide corresponding to human BAG3. | |
100 μL | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction