Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2-associated athanogene 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018172 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-172 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-172 Supplier Abnova Corporation Supplier No. H00009529D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human BAG5 protein.

The protein encoded by this gene is a member of the BAG1-related protein family. BAG1 is an anti-apoptotic protein that functions through interactions with a variety of cell apoptosis and growth related proteins including BCL-2, Raf-protein kinase, steroid hormone receptors, growth factor receptors and members of the heat shock protein 70kDa family. This protein contains a BAG domain near the C-terminus, which could bind and inhibit the chaperone activity of Hsc70/Hsp70. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Sequence: MDMGNQHPSISRLQEIQKEVKSVEQQVIGFSGLSDDKNYKKLERILTKQLFEIDSVDTEGKGDIQQARKRAAQETERLLKELEQNANHPHRIEIQNIFEEAQSLVREKIVPFYNGGNCVTDEFEEGIQDIILRLTHVKTGGKISLRKARYHTLTKICAVQEIIEDCMKKQPSLPLSEDAHPSVAKINFVMCEVNKARGVLIALLMGVNNNETCRHLSCVLSGLIADLDALDVCGRTEIRNYRREVVEDINKLLKYLDLEEEADTTKAFDLRQNHSILKIEKVLKRMREIKNELLQAQNPSELYLSSKTELQGLIGQLDEVSLEKNPCIREARRRAVIEVQTLITYIDLKEALEKRKLFACEEHPSHKAVWNVLGNLSEIQGEVLSFDGNRTDKNYIRLEELLTKQLLALDAVDPQGEEKCKAARKQAVRLAQNILSYLDLKSDEWEY

Specifications

Antigen BCL2-associated athanogene 5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human BAG5 protein.
Formulation PBS with no preservative; pH 7.4
Gene BAG5
Gene Accession No. NM_004873
Gene Alias BAG-5
Gene Symbols BAG5
Host Species Rabbit
Immunogen BAG5 (NP_004864.1, 1 a.a. ∼ 447 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9529
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.