Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI2, Mouse, Clone: 6A12, Abnova™

Catalog No. 89016292 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-292 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-292 Supplier Abnova Corporation Supplier No. H00000576M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BAI2.

BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis. [provided by RefSeq

Sequence: DPAPSACSALASGVLYGAFSLQDLFPTIASGCSWTLENPDPTKYSLYLRFNRQEQVCAHFAPRLLPLDHYLVNFTCLRPSPEEAVAQ

Specifications

Antigen BAI2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BAI2.
Formulation PBS with no preservative; pH 7.4
Gene BAI2
Gene Accession No. NM_001703
Gene Symbols BAI2
Host Species Mouse
Immunogen BAI2 (NP_001694, 22 a.a. ∼ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 576
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.