Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Bak Clone: TC-100,

Catalog No. AM03100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AM03100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM03100UG Supplier MilliporeSigma™ Supplier No. AM03100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Bak Clone: TC-100 in Mouse, Rat samples, and it is validated for Immunoblotting, Immunocytochemistry, Western Blotting

Bak (for bcl-2 homologous antagonist/killer) is a member of the bcl-2 family of proteins. Bak was independently identified by three different groups using either the two hybrid system for identifying adenovirus E1B 19K interacting proteins or by PCR cloning using degenerate BH1 and BH2 domain primers derived from the human bcl-2 gene. Bak shows a 53% homology with bcl-2 in the BH1 and BH2 domains but an overall homology of 28% with bcl-2. The presence of a hydrophobic tail on the protein suggests that, like bcl-2, it is membrane localized. The Bak protein is translated from a 2.2 kb mRNA to yield a 211-216 amino acid protein with a predicted molecular weight of 23.4kDa but migrates by gel electrophoresis as an approximately 30kDa protein under denaturing conditions. The bak gene (bak-1) has been mapped to chromosome 6 and spans approximately 6 kb of DNA. Southern analysis has indicated the presence of two additional bak genes: bak-2 has 97% homology with bak-1 and maps to chromosome 20, and bak-3, a pseudogene, which maps to chromosome 11. Functionally, bak is similar to bax in that it induces apoptosis. The protein pairs with bcl-xL but unlike its pairing partner, has been found in most tissues tested. While conflicting reports exist for the heterodimerization of bak with other members of the bcl-2 family of proteins, it is clear that bak does partner with the E1B 19K protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bak
Applications Immunoblot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone TC-100
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen recombinant human Bak from which the C-terminal and transmembrane domains have been deleted
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.