Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bak Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06536MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-536-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-536-MI Supplier MilliporeSigma Supplier No. 06536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Bak using this Anti-Bak Antibody, NT validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bak
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in storage buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide. Frozen liquid.
Gene Symbols BAK1; BAK; MGC3887; BCL2L7; BAK-LIKE; OTTHUMP00000039620; CDN1; MGC117255
Host Species Rabbit
Immunogen Synthetic peptide (SEEQVAQQDTEEVFRSC) corresponding to amino acid residues 23-38 of human Bak with a cysteine residue added to the C-terminus for conjugation to KLH.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001188
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.