Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAL Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB10618MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10618MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10618MI Supplier MilliporeSigma Supplier No. AB10618
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-BAL Antibody is an antibody against BAL for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation This polyclonal antibody is supplied in 400μL of phosphate buffered saline (pH 7.4) containing 0.1% sodium azide.
Gene Symbols PARP9; DKFZp666B0810; DKFZp686M15238; PARP-9; BAL1; BAL; FLJ26637; Poly; FLJ41418; MGC:7868
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the short isoform of human BAL protein (B-aggressive lymphoma, poly (ADP-ribose) polymerase family member 9, PARP9).
Purification Method Immunoaffinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_031458.1
Target Species Human
Content And Storage −20°C in undiluted aliquotes for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.