Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BARD1, Mouse, Clone: 2A11, Abnova™

Catalog No. 89016293 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-293 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-293 Supplier Abnova Corporation Supplier No. H00000580M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BARD1.

This gene encodes a protein which interacts with the N-terminal region of BRCA1. In addition to its ability to bind BRCA1 in vivo and in vitro, it shares homology with the 2 most conserved regions of BRCA1: the N-terminal RING motif and the C-terminal BRCT domain. The RING motif is a cysteine-rich sequence found in a variety of proteins that regulate cell growth, including the products of tumor suppressor genes and dominant protooncogenes. This protein also contains 3 tandem ankyrin repeats. The BARD1/BRCA1 interaction is disrupted by tumorigenic amino acid substitutions in BRCA1, implying that the formation of a stable complex between these proteins may be an essential aspect of BRCA1 tumor suppression. This protein may be the target of oncogenic mutations in breast or ovarian cancer. [provided by RefSeq

Sequence: RRSRLNREQLLPKLFDGCYFYLWGTFKHHPKDNLIKLVTAGGGQILSRKPKPDSDVTQTINTVAYHARPDSDQRFCTQYIIYEDLCNYHPERVRQGKVWK

Specifications

Antigen BARD1
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 2A11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BARD1.
Formulation PBS with no preservative; pH 7.4
Gene BARD1
Gene Accession No. NM_000465
Gene Symbols BARD1
Host Species Mouse
Immunogen BARD1 (NP_000456, 658 a.a. ∼ 757 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 580
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.