Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BARX homeobox 1, Mouse, Clone: 1E7, Abnova™

Catalog No. 89001319 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-319 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-319 Supplier Abnova Corporation Supplier No. H00056033M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant BARX1.

This gene encodes a member of the Bar subclass of homeobox transcription factors. Studies of the mouse and chick homolog suggest the encoded protein may play a role in developing teeth and craniofacial mesenchyme of neural crest origin. The protein may also be associated with differentiation of stomach epithelia. [provided by RefSeq

Sequence: MGLEKRFEKQKYLSTPDRIDLAESLGLSQLQVKTWYQNRRMKWKKIVLQGGGLESPTKPKGRPKKNSIPTSEQLTEQERAKDAEKPAEVPGEPSDRSRED

Specifications

Antigen BARX homeobox 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant BARX1.
Formulation PBS with no preservative; pH 7.4
Gene BARX1
Gene Accession No. BC009458
Gene Symbols BARX1
Host Species Mouse
Immunogen BARX1 (AAH09458, 1 a.a. ∼ 100 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 56033
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.