Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basophils Mouse anti-Human, Unlabeled, Clone: BSP-1, BD

Catalog No. BDB552754 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552754 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552754 Supplier BD Biosciences Supplier No. 552754

Mouse Monoclonal Antibody

Bsp-1 reacts with a 45 kDa surface antigen on the human erythroleukemic (HEL) cell line, the KU812 basophilic cell line, and peripheral blood basophils, but not on neutrophils or eosinophils. Bsp-1+ basophils also express CD11 and CD13, but lack lymphoid and other granulocytic/monocytic cell surface antigens. However, they express CD45 and HLA-ABC antigens. In peripheral blood, Bsp-1 reacts with a subset of the CD123+/HLA-DR- population. Bsp-1 is a useful tool in the study of basophil function and differentiation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Basophils
Applications Flow Cytometry
Classification Monoclonal
Clone BSP-1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.