Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-B associated transcript 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016938 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-938 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-938 Supplier Abnova Corporation Supplier No. H00007917B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human BAT3 protein.

This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MEPNDSTSTAVEEPDSLEVLVKTLDSQTRTFIVGAQMNVKEFKEHIAASVSIPSEKQRLIYQGRVLQDDKKLQEYNVGGKVIHLVERAPPQTHLPSGASSGTGSASATHGGGSPPGTRGPGASVHDRNANSYVMVGTFNLPSDGSAVDVHINMEQAPIQSEPRVRLVMAQHMIRDIQTLLSRMECRGGPQPQHSQPPPQPPAVTPEPVALSSQTSEPVESEAPPREPMEAEEVEERAPAQNPELTPGPAPAGPTPAPETNAPNHPSPAEYVEVLQELQRLESRLQPFLQRYYEVLGAAATTDYNNNHEGREEDQRLINLVGESLRLLGNTFVALSDLRCNLACTPPRHLHVVRPMSHYTTPMVLQQAAIPIQINVGTTVTMTGNGTRPPPTPNAEAPPPGPGQASSVAPSSTNVESSAEGAPPPGPAPPPATSHPRVIRISHQSVEPVVMMHMNIQDSGTQPGGVPSAPTGPLGPPGHGQTLGQQVPGFPTAPTRVVIARPTPPQARPSHPGGPPVSGTLQGAGLGTNASLAQMVSGLVGQLLMQPVLVAQGTPGMAPPPAPATASASAGTTNTATTAGPAPGGPAQPPPTPQPSMADLQFSQLLGNLLGPAGPGAGGSGVASPTITVAMPGVPAFLQGMTDFLQATQTAPPPPPPPPPPPPAPEQQTMPPPGSPSGGAGSPGGLGLESLSPEFFTSVVQGVLSSLLGSLGARAGSSESIAAFIQRLSGSSNIFEPGADGALGFFGALLSLLCQNFSMVDVVMLLHGHFQPLQRLQPQLRSFFHQHYLGGQEPTPSNIRMATHTLITGLEEYVRESFSLVQVQPGVDIIRTNLEFLQEQFNSIAAHVLHCTDSGFGARLLELCNQGLFECLALNLHCLGGQQMELAAVINGRIRRMSRGVNPSLVSWLTTMMGLRLQVVLEHMPVGPDAILRYVRRVGDPPQPLPEEPMEVQGAERASPEPQRENASPAPGTTAEEAMSRGPPPAPEGGSRDEQDGASAETEPWAAAVPPEWVPIIQQDIQSQRKVKPQPPLSDAYLSGMPAKRRKTMQGEGPQLLLSEAVSRAAKAAGARPLTSPESLSRDLEAPEVQESYRQQLRSDIQKRLQEDPNYSPQRFPNAQRAFADDP

Specifications

Antigen HLA-B associated transcript 3
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human BAT3 protein.
Formulation PBS with no preservative; pH 7.4
Gene BAT3
Gene Accession No. NM_080702.1
Gene Alias BAG-6/BAG6/D6S52E/G3
Gene Symbols BAT3
Host Species Mouse
Immunogen BAT3 (NP_542433.1, 1 a.a. ∼ 1126 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7917
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.