Learn More
basic leucine zipper transcription factor, ATF-like, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events. [provided by RefSeq
Specifications
Specifications
| Antigen | basic leucine zipper transcription factor, ATF-like |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human BATF protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | BATF |
| Gene Accession No. | NM_006399.2 |
| Gene Alias | B-ATF/BATF1/SFA-2/SFA2 |
| Gene Symbols | BATF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.