Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BATF Mouse, PE, Clone: S39-1060, BD

Catalog No. BDB564503 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564503 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564503 Supplier BD Biosciences Supplier No. 564503
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S39-1060 monoclonal antibody specifically binds to BATF. BATF is also known as B-cell-activating transcription factor, Activating transcription factor B, Basic leucine zipper transcription factor ATF-like, and SF-HT-activated gene 2 protein (SFA-2). BATF is a 14 kDa basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. It is upregulated in activated T cells when compared to naοve T and B cells. BATF functions by dimerizing with members of the Jun transcription factor family and binds to AP-1 DNA sequences. BATF plays roles in the differentiation of Th9 cells, Th17 cells, T follicular helper cells, CD8+ effector T cells, CD8+ dendritic cells, Ig isotype-switching B cells, and possibly Th2 cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BATF
Applications Flow Cytometry
Classification Monoclonal
Clone S39-1060
Conjugate PE
Description Basic leucine zipper transcriptional factor ATF-like, SFA-2, B-ATF, Batf
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human BATF Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.