Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB2930MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2930MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2930MI Supplier MilliporeSigma Supplier No. AB2930

Rabbit Polyclonal Antibody

Detect Bax using this Anti-Bax Antibody, a.a. 43-61 hBax validated for use in western blot, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ba,X a.a. 43-61 hBax
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Whole serum without preservatives.
Gene Symbols BAX
Host Species Rabbit
Immunogen Peptide against human Bax aa 43-61
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004324.3; NM_138761.2; NM_138763.2; NM_138764.2; NM_138765.2
Target Species Human
Content And Storage -20°C to -80°C for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.