Learn More
Bax Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
$286.00 - $528.50
Specifications
Antigen | Bax |
---|---|
Clone | 3 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Proper development and maintenance of homeostasis within tissues is achieved through cell proliferation, differentiation, and apoptosis. Members of the Bcl-2 protein family function to either inhibit (Bcl-2 and Bcl-XL) or promote (Bax and Bak) apoptosis.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Bax | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
3 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human | |
Human Bax-α aa. 55-178 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.