Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax Mouse, Unlabeled, Clone: 6A7, BD

Catalog No. BDB556467 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556467 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556467 Supplier BD Biosciences Supplier No. 556467
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Bcl-2 family members are involved in mediating programmed cell death or apoptosis, and share two highly conserved functional regions, Bcl-2 homology 1 and 2 (BH1 and BH2). Several of the family members including Bcl-2 act as inhibitors of apoptosis, whereas others such as Bax promote cell death. It is thought that protein-protein interactions between Bcl-2 family members play an important role in their function. For example, Bax may form homodimers or heterodimers with either Bcl-2 and Bcl-XL (long). Bax homodimers is thought to promote cell death, whereas Bax heterodimerization with either Bcl-2 and Bcl-XL appears to block cell death. When Bax is present in excess, it can counteract the ability of Bcl-2 to inhibit cell death. Bax expression has been identified in a variety of tissues, including lung, stomach, kidney, thymus, lymph nodes, bone marrow, spleen, heart, exocrine pancreas, and brain. Liver, kidney, and the exocrine pancreas have been found to contain little or no Bcl-2. The Bax gene potentially encodes three different proteins Bax-α (21 kDa), Bax-β (24 kDa) and Bax-γ (5 kDa). Bax-α is the most common transcript.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bax
Applications Western Blot
Classification Monoclonal
Clone 6A7
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen N-terminal peptide sequence
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.