Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax (NT) Rabbit anti-Human, Bovine, Feline, Chimpanzee, Canine, Polyclonal, MilliporeSigma™

Catalog No. ABC11MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC11MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC11MI Supplier MilliporeSigma Supplier No. ABC11
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Bax Antibody is an antibody against Bax (N-terminus) for use in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bax (NT)
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols BAX; BCL2L4; Bcl2-L-4
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human BAX at and around the N-terminus.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_620119
Target Species Bovine, Canine, Chimpanzee, Feline, Human
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.