Learn More
Bardet-Biedl syndrome 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant BBS1.
Supplier: Abnova Corporation H00000582A01
Description
Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development. [provided by RefSeq
Sequence: NQHKSNSIKRQTVITTMTTLKKNLADEDAVSCLVLGTENKELLVLDPEAFTILAKMSLPSVPVFLEVSGQFDVEFRLAAACRNGNIYILRRDSKHPKYCIELSAQPVGLSpecifications
Bardet-Biedl syndrome 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant BBS1. | |
BBS1 | |
BBS2L2/FLJ23590/MGC126183/MGC126184/MGC51114 | |
Mouse | |
50 μL | |
Yes | |
582 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_024649 | |
BBS1 | |
BBS1 (NP_078925, 187 a.a. ∼ 295 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.