Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bardet-Biedl syndrome 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016728 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-728 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-728 Supplier Abnova Corporation Supplier No. H00000583B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human BBS2 protein.

This gene is a member of the Bardet-Biedl syndrome (BBS) gene family. Bardet-Biedl syndrome is an autosomal recessive disorder characterized by severe pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation. The proteins encoded by BBS gene family members are structurally diverse and the similar phenotypes exhibited by mutations in BBS gene family members is likely due to their shared roles in cilia formation and function. Many BBS proteins localize to the basal bodies, ciliary axonemes, and pericentriolar regions of cells. BBS proteins may also be involved in intracellular trafficking via microtubule-related transport. The protein encoded by this gene forms a multiprotein BBSome complex with six other BBS proteins

Sequence: MLLPVFTLKLRHKISPRMVAIGRYDGTHPCLAAATQTGKVFIHNPHTRNQHVSASRVFQSPLESDVSLLNINQAVSCLTAGVLNPELGYDALLVGTQTNLLAYDVYNNSDLFYREVADGANVVVLGTLGDISSPLAIIGGNCALQGFNHEGSDLFWTVTGDNVNSLALCDFDGDGKKELLVGSEDFDIRVFKEDEIVAEMTETEIVTSLCPMYGSRFGYALSNGTVGVYDKTSRYWRIKSKNHAMSIHAFDLNSDGVNELITGWSNGKVDARSDRTGEVIFKDNFSSAIAGVVEGDYRMDGHIQLICCSVDGEIRGYLPGTAEMRGNLMDTSAEQDLIRELSQKKQNLLLELRNYEENAKAELASPLNEADGHRGIIPANTRLHTTLSVSLGNETQTAHTELRISTSNDTIIRAVLIFAEGIFTGESHVVHPSIHNLSSSICIPIVPPKDVPVDLHLKAFVGYRSSTQFHVFESTRQLPRFSMYALTSLDPASEPISYVNFTIAERAQRVVVWLGQNFLLPEDTHIQNAPFQVCFTSLRNGGHLHIKIKLSGEITINTDDIDLAGDIIQSMASFFAIEDLQVEADFPVYFEELRKVLVKVDEYHSVHQKLSADMADHSNLIRSLLVGAEDARLMRDMKTMKSRYMELYDLNRDLLNGYKIRCNNHTELLGNLKAVNQAIQRAGRLRVGKPKNQVITACRDAIRSNNINTLFKIMRVGTASS

Specifications

Antigen Bardet-Biedl syndrome 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human BBS2 protein.
Formulation PBS with no preservative; pH 7.4
Gene BBS2
Gene Accession No. BC014140.2
Gene Alias BBS/MGC20703
Gene Symbols BBS2
Host Species Mouse
Immunogen BBS2 (AAH14140.1, 1 a.a. ∼ 721 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 583
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.