Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bardet-Biedl syndrome 7, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018512 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-512 Supplier Abnova Corporation Supplier No. H00055212D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human BBS7 protein.

Mutations in this gene have been observed in patients with Bardet-Biedl syndrome type 7. The encoded protein may play a role in eye, limb, cardiac and reproductive system development. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MDLILNRMDYLQVGVTSQKTMKLIPASRHRATQKVVIGDHDGVVMCFGMKKGEAAAVFKTLPGPKIARLELGGVINTPQEKIFIAAASEIRGFTKRGKQFLSFETNLTESIKAMHISGSDLFLSASYIYNHYCDCKDQHYYLSGDKINDVICLPVERLSRITPVLACQDRVLRVLQGSDVMYAVEVPGPPTVLALHNGNGGDSGEDLLFGTSDGKLALIQITTSKPVRKWEIQNEKKRGGILCIDSFDIVGDGVKDLLVGRDDGMVEVYSFDNANEPVLRFDQMLSESVTSIQGGCVGKDSYDEIVVSTYSGWVTGLTTEPIHKESGPGEELKINQEMQNKISSLRNELEHLQYKVLQERENYQQSSQSSKAKSAVPSFGINDKFTLNKDDASYSLILEVQTAIDNVLIQSDVPIDLLDVDKNSAVVSFSSCDSESNDNFLLATYRCQADTTRLELKIRSIEGQYGTLQAYVTPRIQPKTCQVRQYHIKPLSLHQRTHFIDHDRPMNTLTLTGQFSFAEVHSWVVFCLPEVPEKPPAGECVTFYFQNTFLDTQLESTYRKGEGVFKSDNISTISILKDVLSKEATKRKINLNISYEINEVSVKHTLKLIHPKLEYQLLLAKKVQLIDALKELQIHEGNTNFLIPEYHCILEEADHLQEEYKKQPAHLERLYGMITDLFIDKFKFKGTNVKTKVPLLLEILDSYDQNALISFFDAA

Specifications

Antigen Bardet-Biedl syndrome 7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human BBS7 protein.
Formulation PBS with no preservative; pH 7.4
Gene BBS7
Gene Accession No. NM_176824.1
Gene Alias BBS2L1/FLJ10715
Gene Symbols BBS7
Host Species Rabbit
Immunogen BBS7 (NP_789794.1, 1 a.a. ∼ 715 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55212
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.