Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BCKDHA, Polyclonal, Abnova

Catalog No. 89004967 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-967 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-967 Supplier Abnova Corporation Supplier No. H00000593B01

Mouse Polyclonal

The second major step in the catabolism of the branched-chain amino acids, isoleucine, leucine, and valine, is catalyzed by the branched-chain alpha-keto acid dehydrogenase complex (BCKD; EC 1.2.4.4), an inner-mitochondrial enzyme complex that consists of 3 catalytic components: a heterotetrameric (alpha2, beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase (E2; MIM 248610), and a homodimeric dihydrolipoamide dehydrogenase (E3; MIM 238331). The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase. The BCKDHA gene encodes the alpha subunit of E1, and the BCKDHB gene (MIM 248611) encodes the beta subunit of E1.

Detection Antibody, Western Blotting

Specifications

Antigen branched chain keto acid dehydrogenase E1, alpha polypeptide
Classification Polyclonal
Conjugate Unconjugated
Host Species Mouse
Immunogen BCKDHA (NP_000700.1, 1 a.a. ∽445 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Target Species Human
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.