Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BCL-2α Ab-4, Clone: 10C4, Epredia™ LabVision

Catalog No. MS598B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
500 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS598B0 100 μL
MS598B1 500 μL
MS598B 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS598B0 Supplier Epredia Supplier No. MS598B0

Mouse Monoclonal Antibody

Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Host Species: Mouse
Clone: 10C4
Isotype: IgG1
Cross Reactivity: Mouse and Rat. Does not react with human. Others-not tested.
Epitope: aa 61-76
Immunogen: A Synthetic peptide, aa 61-76 (Cys-VHREMAARTSPLRPLV), of Mouse bcl-2 protein.
Molecular Weight: 25-26kDa
Positive Control: MAD109 cells.
Cellular Localization: Cytoplasm

Ab-4 is particularly designed for research on mouse and rat bcl-2alpha protein. It does not react with human bcl-2alpha protein.

Immunofluorescence, Immunoprecipitation, Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen BCL-2α Ab-4
Classification Monoclonal
Concentration 200μg/mL
Host Species Mouse
Immunogen A Synthetic peptide, aa 61-76 (Cys-VHREMAARTSPLRPLV), of Mouse bcl-2 protein.
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Isotype IgG1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.