Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Mouse anti-Human, Alexa Fluor 647, Clone: N46-467, BD

Catalog No. BDB562531 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562531 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562531 Supplier BD Biosciences Supplier No. 562531

Mouse Monoclonal Antibody

The N46-467 monoclonal antibody specifically binds to Bcl-2 (pS70), ie, the Bcl-2 protein phosphorylated at the Ser70 site. Bcl-2 is a ∽ 26 kDa intracellular, integral membrane protein found primarily in the nuclear envelope, endoplasmic reticulum and outer mitochondrial membrane. Bcl-2 is encoded by the BCL2 (B-cell CLL/lymphoma 2) gene and is also known as Apoptosis regulator Bcl-2. Members of the Bcl-2 family play a major role in regulating the response of cells to apoptotic signals. Bcl-2 is one of the anti-apoptotic members of the Bcl-2 family. Bcl-2 knockout mice showed pronounced lymphoid apoptosis and other apoptosis related lesions later in life. Bcl-2 is a proto-oncogene because it blocks apoptosis and provides a selective survival advantage in many cell types and thus contributes to tumorigenesis. It has been implicated in several types of cancers, such as breast, prostate, and melanoma . Bcl-2 contains multiple phosphorylation sites including Thr56, Ser70, Thr74 and Ser87. Phosphorylation of Bcl-2 Ser70 has been shown to be a mitotic marker. Phosphorylation at this site regulates Bcl-2's anti-apoptotic activity and has recently been implicated in promoting autophagy. Several studies have shown that Bcl-2 phosphorylation is caused by c-Jun N-terminal kinase (JNK).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Flow Cytometry
Classification Monoclonal
Clone N46-467
Conjugate Alexa Fluor 647
Description BCL2; Apoptosis regulator Bcl-2; B-cell CLL/lymphoma 2; PPP1R50
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.