Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl2 Mouse anti-Human, Canine, Feline, Clone: 100, MilliporeSigma™

Catalog No. 05729MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-729-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-729-MI Supplier MilliporeSigma Supplier No. 05729
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Bcl2 using this Anti-Bcl2 Antibody, clone 100 validated for use in flow cytometry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl2
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 100
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 70% storage buffer PBS with 0.1% sodium azide and 30% glycerol. Store at −20deg;C.
Gene Accession No. NM_000633
Gene Symbols BCL2, Bcl-2
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 41-54 of human Bcl2 protein. The immunizing sequence is identical in canine and feline.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Feline, Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.