Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Mouse, Unlabeled, Clone: 7, BD

Catalog No. BDB610539 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610539 150 μg
BDB610538 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610539 Supplier BD Biosciences Supplier No. 610539
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human hematopoietic malignancies are often characterized by gene rearrangements which deregulate gene expression. The Bcl-2 gene (B-cell leukemia/lymphoma 2) was discovered by walking the chromosomal breakpoints t(14,18). This gene consists of two exons and several transcripts that encode two different protein products, Bcl-2α and Bcl-2β. Both are approximately 26-30kDa and differ at their C-terminal region. Bcl-2 suppresses programmed cell death (apoptosis) and this property appears to be modulated by homodimerization or association with Bax and Bad, two cell death promoters.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Western Blot
Classification Monoclonal
Clone 7
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.