Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Mouse anti-Human, Clone: AW604, MilliporeSigma™

Catalog No. 05826MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-826-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-826-MI Supplier MilliporeSigma Supplier No. 05826
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Bcl-2 Antibody, clone AW604 is validated for use in immunoprecipitation, western blotting for the detection of Bcl-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AW604
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_000633
Gene Symbols BCL2, Bcl-2
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 41-54 (C-GAAPAPGIFSSQPG) of human Bcl-2
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.