Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 with control Mouse anti-Human, Unlabeled, Clone: Bcl-2/100, BD

Catalog No. BDB551107 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551107 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551107 Supplier BD Biosciences Supplier No. 551107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl-2 is considered to be novel among proto-oncogenes because it blocks apoptosis (programmed cell death) in many cell types. Apoptosis is an active form of cellular suicide that typically requires new RNA and protein synthesis and is associated with distinct morphological changes including cell shrinkage, cytoplasm membrane blebbing, nuclear fragmentation and DNA degradation. Because Bcl-2 blocks apoptosis it may contribute to tumorigenisis by prolonging cell survival, rather than by accelerating the rate of cell proliferation. Human Bcl-2 protein migrates at a molecular weight of ∽26 kDa by SDS-PAGE. Bcl-2/100 recognizes a 26 kDa band representing human Bcl-2. Additional minor bands at 27-31 kDa and 18-21 kDa may also be visualized. The 27-31 kDa upper band may represent a larger isoform, whereas the 18-21 kDa lower band may be an internal translation or proteolytic product, therefore, a synthetic peptide corresponding to amino acids 41-54 (GAAPAPGIFSSQPG) of human Bcl-2 was used as immunogen. This peptide sequence is not conserved between human and mouse. Bcl-2/100 does not cross-react with mouse Bcl-2. For detection of mouse Bcl-2 refer to clone 3F11 (Cat. No. 554218), polyclonal rabbit anti-rat/mouse Bcl-2 antiserum (Cat. No. 554087), and polyclonal rabbit anti-mouse Bcl-2 antiserum (Cat. No. 554279).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2 with control
Applications Western Blot
Classification Monoclonal
Clone Bcl-2/100
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Includes Purified Mouse Anti-Human Bcl-2 (50μg); Jurkat Cell Lysate (50μg)
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.