Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Mouse anti-Human, PE, Clone: BCL-2/100, BD

Catalog No. BDB340576 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340576 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340576 Supplier BD Biosciences Supplier No. 340576
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-bcl-2, clone 100, is derived from the fusion of Sp2/0-Ag14 cells with spleen cells from BALB/c mice immunized with the bcl-2α peptide (amino acids 41-54).

Anti–bcl-2 recognizes a 26-kilodalton (kd) protein encoded by the 230 kb bcl-2 protooncogene. The protein is a regulator of the apoptotic process, and elevated levels can provide resistance to cell death. The bcl-2 protein is expressed in the interior of cells, frequently localized to the mitochondrial membrane.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone BCL-2/100
Concentration 1μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human Bcl-2 synthetic peptide aa. 41-54
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.