Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Hamster anti-Mouse, FITC, Clone: 3F11, BD

Catalog No. BDB554221 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554221 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554221 Supplier BD Biosciences Supplier No. 554221
Add to Cart
Add to Cart

Includes

FITC Hamster Anti-Mouse Bcl-2 and FITC Hamster IgG Isotype Control (Anti-Trinitrophenol (TNP))

Hamster Monoclonal Antibody

Programmed cell death (apoptosis) is a normal physiologic process which occurs during embryonic development as well as in maintenence of tissue homeostasis. The apoptotic program is characterized by certain morphological features. These include changes in the plasma membrane such as loss of membrane asymmetry and attachment, a condensation of the cytoplasm and nucleus, and internucleosomal cleavage of DNA. In the final stages, the dying cells become fragmented into “apoptotic bodies” which are rapidly eliminated by phagocytic cells without eliciting significant inflammatory damage to surrounding cells. Members of the Bcl-2 family play a major role in regulating the cellular response to apoptotic signals. Bcl-2 is considered to be a novel proto-oncogene because it blocks apoptosis in many cell types. Bcl-2 is thought to provide selective survival advantage for cells by blocking apoptosis and thus may contribute to tumorigenesis. Bcl-2 is a 26 kD intracellular, integral membrane protein found primarily in the nuclear envelope, endoplasmic reticulum and outer mitochondrial membrane.

Clone 3F11 reacts with mouse Bcl-2. It does not cross-react with human Bcl-2. Gel purified recombinant mouse Bcl-2 protein was used as immunogen.

Clone A19-3 is an Armenian hamster IgG isotype control which is specific for the hapten trinitrophenol (TNP). This hapten is not expressed on human cells or human cell lines. The solutions are free of unconjugated antibody. The 3F11 and A19-3 FITC conjugates are matched in F/P ratios. The optimum F/P ratio was experimentally determined by flow cytometric analysis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Flow Cytometry
Classification Monoclonal
Clone 3F11
Conjugate FITC
Host Species Hamster
Immunogen Recombinant Mouse Bcl-2, TNP-keyhole limpet hemocyanin
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.