Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Bcl-W, Clone: 16H12,

Catalog No. MAB17002MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB17002MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB17002MI Supplier MilliporeSigma™ Supplier No. MAB17002

Specifications

Antigen Bcl-W
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 16H12
Conjugate Unconjugated
Formulation Purified antibody. Liquid, at a concentration of 1mg/mL in PBS, pH 7.4, containing 0.05% Bronidox as a preservative.
Gene Accession No. NM_004050.3
Gene Symbols BCL2L2, KIAA0271, BCLW, BCL-W
Host Species Rat
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.