Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-X set Mouse, Unlabeled, Clone: 2H12, BD

Catalog No. BDB551022 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551022 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551022 Supplier BD Biosciences Supplier No. 551022
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the Bcl-2 family play a major role in regulating the response of cells to a wide variety of apoptotic signals. Some Bcl-2 family members like Bcl-2, block apoptosis, whereas others promote apoptosis and inhibit Bcl-2 activity. Bcl-x is a Bcl-2 homologue that has two isoforms, resulting from alternative splicing. Bcl-xL (long) is a 241 amino acid protein (25-29 kDa) that is 47% homologous to Bcl-2 on the amino acid level. Bcl-xS (short) is a 178 amino acid protein (∼ 19.5 kDa) lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 protein family. Bcl-xL blocks cell death, whereas Bcl-xS inhibits Bcl-2 and promotes cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-X set
Applications Western Blot
Classification Monoclonal
Clone 2H12
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.