Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-X Mouse, Unlabeled, Clone: 4, BD

Catalog No. BDB610210 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610210 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610210 Supplier BD Biosciences Supplier No. 610210

Mouse Monoclonal Antibody

Bcl-x is related to the Bcl-2 protein and can function independently of Bcl-2 in regulating apoptosis (programmed cell death). The bcl-x transcript is found in a number of tissues, with the highest levels in the lymphoid and central nervous systems. Two distinct cDNA species, bcl-xL and bcl-xS, have been observed and appear to arise from alternate 5′ splice sites located within the first coding exon of the bcl-x gene. Bcl-xL is composed of 233 amino acids and is similar in size and structure to Bcl-2. Bcl-xS lacks 63 amino acids corresponding to the region of Bcl-xL with the highest degree of homology to Bcl-2. Like Bcl-2, Bcl-xL has been reported to inhibit cell death upon growth factor withdrawal in an IL-3-dependent cell line. However, Bcl-xS inhibits the ability of Bcl-2 to enhance cell survival in the absence of growth factors.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-X
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.