Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-XL and BclXs Mouse anti-Human, Clone: 7B2.5, MilliporeSigma™

Catalog No. MAB3121MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3121MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3121MI Supplier MilliporeSigma Supplier No. MAB3121
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Bcl-XL and BclXs using this Anti-Bcl-XL and BclXs Antibody, clone 7B2.5 validated for use in flow cytometry, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-XL and BclXs
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7B2.5
Conjugate Unconjugated
Formulation Purified immunoglobulin - Ig fraction. Liquid in phosphate buffer saline with 0.1% sodium azide.
Gene Accession No. NM_001191.2; NM_138578.1
Gene Symbols BCL2L1, BCLX, bcl-xL, DKFZp781P2092, BCL-XL/S, Bcl-X, BCL2L, bcl-xS
Host Species Mouse
Immunogen Recombinant Bcl-XS.
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.